Learn More
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III), 99%
Vapor deposition | CAS: 14877-41-9 | C33H57CoO6 | 608.75 g/mol
$120.04 - $1207.47
Chemical Identifiers
| CAS | 14877-41-9 |
|---|---|
| Molecular Formula | C33H57CoO6 |
| Molecular Weight (g/mol) | 608.75 |
| MDL Number | MFCD00233617,MFCD00233617 |
| InChI Key | UJRJNARBHKKZST-UHFFFAOYSA-K |
| Synonym | cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
| PubChem CID | 131675252 |
| SMILES | [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4399302
|
Thermo Scientific Chemicals
04399302 |
0.5 g |
Each for $120.04
|
|
|||||
|
AA4399304
|
Thermo Scientific Chemicals
04399304 |
2 g |
Each for $365.42
|
|
|||||
|
AA4399309
|
Thermo Scientific Chemicals
04399309 |
10 g |
Each for $1,207.47
|
|
|||||
Description
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III) is used precursor for aerosol-assisted CVD of mixed-conducting ceramic films of Sr-Co-Fe perovskite phases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14877-41-9 | |
| 608.75 | |
| UJRJNARBHKKZST-UHFFFAOYSA-K | |
| 131675252 |
| C33H57CoO6 | |
| MFCD00233617,MFCD00233617 | |
| cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
| [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C |
Specifications
| 14877-41-9 | |
| Green | |
| C33H57CoO6 | |
| 0.5 g | |
| Insoluble in water. | |
| [Co+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C | |
| 608.75 | |
| 608.72 | |
| Powder |
| 254°C to 256°C | |
| 120°C (0.5 mmHg)(sublimation) | |
| MFCD00233617,MFCD00233617 | |
| cobalt tris 2,2,6,6-tetramethyl-3,5-heptanedionate, cobalt 3+ tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
| UJRJNARBHKKZST-UHFFFAOYSA-K | |
| cobalt(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate) | |
| 131675252 | |
| 99% | |
| Tris (2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt (III) |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only