missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2,2,2-trifluoroethyl) Phosphate 96.0+%, TCI America™
Supplier: TCI America T320325G
Specifications
| Tris(2,2,2-trifluoroethyl) Phosphate | |
| -22°C | |
| 40°C | |
| MFCD00233117 | |
| 3272 | |
| ZMQDTYVODWKHNT-UHFFFAOYSA-N | |
| tris(2,2,2-trifluoroethyl) phosphate | |
| 303454 | |
| ≥96.0% (GC) |
| 358-63-4 | |
| Colorless | |
| C6H6F9O4P | |
| 25 g | |
| Phosphoric Acid Tris(2,2,2-trifluoroethyl) Ester | |
| C(C(F)(F)F)OP(=O)(OCC(F)(F)F)OCC(F)(F)F | |
| 344.069 | |
| 344.07 | |
| Liquid |
Chemical Identifiers
| 358-63-4 | |
| 344.069 | |
| ZMQDTYVODWKHNT-UHFFFAOYSA-N | |
| 303454 | |
| C(C(F)(F)F)OP(=O)(OCC(F)(F)F)OCC(F)(F)F |
| C6H6F9O4P | |
| MFCD00233117 | |
| Phosphoric Acid Tris(2,2,2-trifluoroethyl) Ester | |
| tris(2,2,2-trifluoroethyl) phosphate |
Safety and Handling
TSCA : No