missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,2-trifluoroethyl) Phosphate 96.0+%, TCI America™

Catalog No. T32035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32035G 5 g
T320325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32035G Supplier TCI America Supplier No. T32035G
Only null left

Chemical Identifiers

CAS 358-63-4
Molecular Formula C6H6F9O4P
Molecular Weight (g/mol) 344.069
MDL Number MFCD00233117
InChI Key ZMQDTYVODWKHNT-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(2,2,2-trifluoroethyl) Ester
PubChem CID 303454
IUPAC Name tris(2,2,2-trifluoroethyl) phosphate
SMILES C(C(F)(F)F)OP(=O)(OCC(F)(F)F)OCC(F)(F)F

Specifications

Melting Point -22°C
Color Colorless
Boiling Point 40°C
Quantity 5 g
UN Number 3272
Formula Weight 344.07
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2,2,2-trifluoroethyl) Phosphate