missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,2-trifluoroethyl) Borate 95.0+%, TCI America™

Catalog No. t30101g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30101G Supplier TCI America Supplier No. T30101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 659-18-7
Molecular Formula C6H6BF9O3
Molecular Weight (g/mol) 307.907
MDL Number MFCD00093768
InChI Key DIEXQJFSUBBIRP-UHFFFAOYSA-N
Synonym Boric Acid Tris(2,2,2-trifluoroethyl) Ester
PubChem CID 3599767
IUPAC Name tris(2,2,2-trifluoroethyl) borate
SMILES B(OCC(F)(F)F)(OCC(F)(F)F)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 125°C
Quantity 1 g
Formula Weight 307.91
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(2,2,2-trifluoroethyl) Borate