missing translation for 'onlineSavingsMsg'
Learn More

Tris(1H-benzotriazol-1-yl)methane 95.0+%, TCI America™

Catalog No. t31411g
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31411G 1 g
T3141200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31411G Supplier TCI America Supplier No. T31411G
Only null left

Chemical Identifiers

CAS 88088-95-3
Molecular Formula C19H13N9
Molecular Weight (g/mol) 367.376
MDL Number MFCD00514776
InChI Key VENLCXAGJRTBFL-UHFFFAOYSA-N
PubChem CID 3733734
IUPAC Name 1-[bis(benzotriazol-1-yl)methyl]benzotriazole
SMILES C1=CC=C2C(=C1)N=NN2C(N3C4=CC=CC=C4N=N3)N5C6=CC=CC=C6N=N5

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 1 g
Formula Weight 367.38
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1H-benzotriazol-1-yl)methane