missing translation for 'onlineSavingsMsg'
Learn More

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97.0+%, TCI America™

Catalog No. p-7137519
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29931G 1 g
T29935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29931G Supplier TCI America Supplier No. T29931G

Chemical Identifiers

CAS 510758-28-8
Molecular Formula C30H30N10
Molecular Weight (g/mol) 530.64
MDL Number MFCD09265124
InChI Key WKGZJBVXZWCZQC-UHFFFAOYSA-N
Synonym tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta
PubChem CID 11203363
IUPAC Name tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
SMILES C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1

Specifications

Melting Point 138°C
Quantity 1 g
Formula Weight 530.64
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
TSCA No
Recommended Storage Refrigerator