missing translation for 'onlineSavingsMsg'
Learn More

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%

Catalog No. p-7057124
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66485MD 250 mg
AAH6648503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66485MD Supplier Thermo Scientific Chemicals Supplier No. H66485MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 510758-28-8 | C30H30N10 | 530.64 g/mol

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

Solubility
Soluble in DMSO, DMF.

Notes
Store at -20°C. Protect from heat. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 510758-28-8
Molecular Formula C30H30N10
Molecular Weight (g/mol) 530.64
MDL Number MFCD09265124
InChI Key WKGZJBVXZWCZQC-UHFFFAOYSA-N
Synonym tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta
PubChem CID 11203363
IUPAC Name 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
SMILES C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1

Specifications

Melting Point 132°C to 143°C
Color White
Quantity 250 mg
Solubility Information Soluble in DMSO,DMF.
Formula Weight 530.63
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only