missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III) 95.0+%, TCI America™

Catalog No. T17355G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17355G 5 g
T17351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17355G Supplier TCI America Supplier No. T17355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17904-83-5
Molecular Formula C57H44EuN2O6
Molecular Weight (g/mol) 1004.951
MDL Number MFCD01321202
InChI Key DYKOLWWJTALFFU-RWBKAWJDSA-N
Synonym eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen
PubChem CID 14205791
IUPAC Name europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline
SMILES C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu]

Specifications

Melting Point 191°C
Quantity 5 g
Formula Weight 1001.93
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)