missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,10-phenanthroline)ruthenium(II) Bis(hexafluorophosphate) 98.0+%, TCI America™

Catalog No. p-7137637
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3208200MG 200 mg
T32081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3208200MG Supplier TCI America Supplier No. T3208200MG
Only null left

Chemical Identifiers

CAS 60804-75-3
Molecular Formula C36H24F12N6P2Ru
Molecular Weight (g/mol) 931.628
InChI Key YRYUXGTVQZIGNQ-UHFFFAOYSA-N
PubChem CID 15198714
IUPAC Name 1,10-phenanthroline;ruthenium(2+);dihexafluorophosphate
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.[Ru+2]

Specifications

Quantity 200 mg
Formula Weight 931.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1,10-phenanthroline)ruthenium(II) Bis(hexafluorophosphate)