missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific™

Catalog No. AC422255000
Change view
Click to view available options
500mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC422255000 500mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422255000 Supplier Thermo Scientific Chemicals Supplier No. 422255000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Indicator

Specifications

CAS 14634-91-4
Density 1.0060g/mL
Packaging Glass bottle
Molecular Formula C36H24FeN6O4S
Quantity 500mL
MDL Number MFCD00036428
Specific Gravity 1.006
Synonym ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]S(=O)(=O)[O-].[Fe+2]
IUPAC Name iron(2+);1,10-phenanthroline;sulfate
Molecular Weight (g/mol) 692.53
PubChem CID 84567
Formula Weight 692.53
Chemical Name or Material Tris(1, 10-phenanthroline)iron(II) sulfate, 0.025M Aqueous Solution
Show More Show Less
EINECSNumber 238-676-6

RUO – Research Use Only