missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate) 97.0+%, TCI America™

Catalog No. p-7137796
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35541G 1 g
T35545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35541G Supplier TCI America Supplier No. T35541G
Only null left

Chemical Identifiers

CAS 28277-59-0
Molecular Formula C36H24CoF18N6P3
Molecular Weight (g/mol) 1034.46
MDL Number MFCD30749311
InChI Key JYGNLHURFRGGRD-UHFFFAOYSA-N
PubChem CID 15494322
IUPAC Name cobalt(3+) tris(1,10-phenanthroline) tris(hexafluoro-λ⁵-phosphanuide)
SMILES [Co+3].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Quantity 1 g
Formula Weight 1034.46
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate)