missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate) 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 1034.46 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris(1,10-phenanthroline)cobalt(III) Tris(hexafluorophosphate) |
Safety and Handling
| TSCA | No |