missing translation for 'onlineSavingsMsg'
Learn More

Tris(1,10-phenanthroline)cobalt(II) Bis(hexafluorophosphate) 98.0+%, TCI America™

Catalog No. p-7137795
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35531G 1 g
T35535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35531G Supplier TCI America Supplier No. T35531G
Only null left

Chemical Identifiers

CAS 31876-74-1
Molecular Formula C36H24CoF12N6P2

Specifications

Quantity 1 g
Formula Weight 889.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(1,10-phenanthroline)cobalt(II) Bis(hexafluorophosphate)