missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1,10-phenanthroline)cobalt(II) Bis(hexafluorophosphate) 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 889.49 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris(1,10-phenanthroline)cobalt(II) Bis(hexafluorophosphate) |
Safety and Handling
| TSCA | No |