missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 32°C |
| Boiling Point | 72°C |
| Quantity | 1 g |
| Formula Weight | 548.06 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |