missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropylene Glycol Diacrylate (stabilized with MEHQ) 90.0+%, TCI America™
$42.87 - $236.73
Chemical Identifiers
| CAS | 42978-66-5 |
|---|---|
| Molecular Formula | C15H24O6 |
| MDL Number | MFCD00064598 |
Chemical Identifiers
| 42978-66-5 | |
| C15H24O6 | |
| MFCD00064598 |
Specifications
| 42978-66-5 | |
| C15H24O6 | |
| 25 g | |
| 300.35 | |
| Liquid |
| Yellow | |
| MFCD00064598 | |
| 3082 | |
| ≥90.0% (GC) | |
| Tripropylene Glycol Diacrylate (stabilized with MEHQ) |
Safety and Handling
RTECSNumber : AT4690000
TSCA : Yes