missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tripropylene Glycol Diacrylate (stabilized with MEHQ) 90.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| UN Number | 3082 |
| Formula Weight | 300.35 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tripropylene Glycol Diacrylate (stabilized with MEHQ) |
Safety and Handling
| RTECSNumber | AT4690000 |
| TSCA | Yes |