missing translation for 'onlineSavingsMsg'
Learn More

Tripropylamine 98.0+%, TCI America™

Catalog No. T052225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T052225ML 25 mL
T0522500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T052225ML Supplier TCI America Supplier No. T052225ML

Chemical Identifiers

CAS 102-69-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00009363
InChI Key YFTHZRPMJXBUME-UHFFFAOYSA-N
Synonym tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
PubChem CID 7616
ChEBI CHEBI:38880
IUPAC Name tripropylamine
SMILES CCCN(CCC)CCC

Specifications

Color Yellow
Boiling Point 156°C
Quantity 25 mL
UN Number 2260
Formula Weight 143.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Tripropylamine
EINECSNumber (2)-3553
RTECSNumber TX1575000
TSCA Yes