missing translation for 'onlineSavingsMsg'
Learn More

Tripropionin, 97%

Catalog No. AC275101000
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275101000 100 g Glass bottle
AC275102500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275101000 Supplier Thermo Scientific Chemicals Supplier No. 275101000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 139-45-7 | C12H20O6 | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139-45-7
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00027005
InChI Key YZWRNSARCRTXDS-UHFFFAOYSA-N
Synonym tripropionin, glycerol tripropionate, glyceryl tripropionate, tripropionine, glyceryl tripropanoate, propionin, tri, glycerine tripropionate, tripropionylglycerol, 1,2,3-propanetriol, tripropanoate, propionic acid triglyceride
PubChem CID 8763
IUPAC Name 2,3-di(propanoyloxy)propyl propanoate
SMILES CCC(=O)OCC(COC(=O)CC)OC(=O)CC

Specifications

Color Colorless
Density 1.0820g/mL
Boiling Point 151°C (2.0 mmHg)
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.434
Quantity 100 g
Specific Gravity 1.082
Formula Weight 260.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tripropionin, 97%
Show More Show Less
Hazard Category LIGHT SENSITIVE
EINECSNumber 205-365-1
RTECSNumber YK6000000
TSCA TSCA

RUO – Research Use Only