missing translation for 'onlineSavingsMsg'
Learn More

Tripropargylamine 98.0+%, TCI America™

Catalog No. p-7137518
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29921G 1 g
T29925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29921G Supplier TCI America Supplier No. T29921G
Only null left

Chemical Identifiers

CAS 6921-29-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00008577
InChI Key ZHOBJWVNWMQMLF-UHFFFAOYSA-N
Synonym tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb
PubChem CID 23351
IUPAC Name N,N-bis(prop-2-ynyl)prop-2-yn-1-amine
SMILES C#CCN(CC#C)CC#C

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 g
UN Number 1993
Formula Weight 131.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Tripropargylamine
RTECSNumber YK7660000
TSCA No
Recommended Storage Refrigerator