missing translation for 'onlineSavingsMsg'
Learn More

Tripropargylamine, 97%

Catalog No. p-7035733
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5345903 1 g
AAH5345906 5 g
AAH5345914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5345903 Supplier Thermo Scientific Chemicals Supplier No. H5345903
Only null left

CAS: 6921-29-5 | C9H9N | 131.178 g/mol

Tripropargylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripropargylamine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6921-29-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00008577
InChI Key ZHOBJWVNWMQMLF-UHFFFAOYSA-N
Synonym tripropargylamine, tri-2-propynylamine, tris propyn-2-yl amine, 2-propyn-1-amine, n,n-di-2-propynyl, unii-5e92gfj1aj, n,n-di-2-propynyl-2-propyn-1-amine, 5e92gfj1aj, n,n-diprop-2-yn-1-ylprop-2-yn-1-amine, tripropargyl amine, acmc-20ajcb
PubChem CID 23351
IUPAC Name N,N-bis(prop-2-ynyl)prop-2-yn-1-amine
SMILES C#CCN(CC#C)CC#C

Specifications

Density 0.927
Boiling Point 79°C to 85°C (11 mmHg)
Flash Point 57°C (135°F)
Refractive Index 1.484
Linear Formula (HC≡CCH2)3N
Quantity 1 g
UN Number UN1993
Formula Weight 131.18
Percent Purity 97%
Chemical Name or Material Tripropargylamine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 230-038-5
RTECSNumber YK7660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only