missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrippNT TRIPLE GLOVE BOX HLDR CLR MAG
Specifications
Specifications
| Color | Clear |
| Material | Acrylic |
| Style | Triple |
| For Use With (Application) | Ideal for labs, clinics, exam rooms, and other industrial settings |
| Includes | Keyhole or magnet mount |