missing translation for 'onlineSavingsMsg'
Learn More

TrippNT DISPENSER APPAREL 3 BINS ACR

Catalog No. 50855985
Encompass
Change view
Click to view available options
Apparel Dispenser
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50855985 Apparel Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50855985 Supplier TrippNT Supplier No. TRP 50127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylic dispenser is high capacity storage for your protective wear. Mounted at entry way, or in lab, just grab and go after these are mounted on the wall or seated on the bench.

Specifications

Color White
Depth (English) 9 in.
Width (English) 20 in.
Height (English) 19 in.
Material PVC, Acrylic
Mounting Keyhole Slots
No. of Compartments 3
Description Large Apparel Dispensing Bin
For Use With (Equipment) Coverall, lab coats, arm protection and more
Includes Dust protecting lid