missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Pipette Storage Carts

Catalog No. NC9984940
Encompass
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
NC9984940 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9984940 Supplier TrippNT Supplier No. TRP 50741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Product Type Angled Bin Cart
Color White
For Use With (Application) Storing pipets, bottles and other items at a secure angle in separate bins to protect while moving
Includes Fifteen 4.6 x 4.6 x 12 in. bins; Two 9.2 x 4.6 x 12 in. bins; Four 3 inch casters, 2 with brakes; Cutaway handles
Material Plastic
Wheel Type 4 x 3 in. rubber casters, 2 with brakes