missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrippNT TRIPLE HEART GLOVE BOX HOLDER
Specifications
Specifications
| Description | Heart Glove Box Holder |
| Color | White |
| Product Type | Heart Glove Box Holder |
| For Use With (Application) | Holds three boxes of gloves |
| Width (English) | 11 in. |