missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Vertical Glove Box/Kimwipe™ Combination Holder

Catalog No. 50856053
Encompass
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
50856053 Keyhole Slots
50856054 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50856053 Supplier TrippNT Supplier No. TRP 50379

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A lean approach for displaying and storing cleaning supplies in laboratories or manufacturing plants. Holds one box of gloves, one box of small Kimwipes™ and one box of large Kimwipes™.

  • A convenient way to store glove boxes and wipers in laboratories or manufacturing plants
  • Regular tension design with open top and closed bottom
  • Smooth finish with no rough, or sharp edges
  • Available with a magnetic or keyhole mount
  • Front dispensing

Specifications

Product Type Combination Holder
Color White
Dimensions (L x W x H) 5.25 x 3.75 x 31.25 in.
Holds Disposable Glove Boxes, Labwipes
Material PVC
Mounting Keyhole Slots
No. of Compartments 1
Product Style Top Loading
For Use With (Application) Storing Cleaning Materials in Laboratories and Manufacturing Facilities
Thickness (English) 0.25 in.