missing translation for 'onlineSavingsMsg'
Learn More

Tripotassium citrate monohydrate, 99+%

Catalog No. AA33222A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33222A3 2 kg
AA3322236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33222A3 Supplier Thermo Scientific Chemicals Supplier No. 033222A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6100-05-6 | C6H7K3O8 | 324.41 g/mol

Tripotassium citrate hydrate is used in food and beverage industries. It finds application as buffering, sequestering or emulsifying agent. It is also used as a potassium source in pharmaceuticals and an active ingredient to treat urinary duct stones. It is added to oral care products to reduce the teeth sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
MDL Number MFCD00150442
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 230°C (decomposition)
Density 1.98 g/mL
Assay Percent Range >99%
Quantity 2 kg
Merck Index 14,7623
Solubility Information Soluble in water and glycerol; Practically insoluble in alcoholSoluble in water and glycerol; Insoluble in alcohol
Formula Weight 324.42 (306.41 Anhydrous)
Percent Purity ≥99%
Physical Form Granules
Chemical Name or Material Tripotassium citrate monohydrate
EINECSNumber 212-755-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only