missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine

Catalog No. OXPO5511D
Change view
Click to view available options
Quantity:
100/Pk.
Format:
55mm Contact Plate
Media Type:
Tryptone Soya Agar
Manufacturing Origin:
Germany
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
OXPO5511D 100/Pk. 55mm Contact Plate Tryptone Soya Agar Germany
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXPO5511D Supplier Thermo Scientific™ Supplier No. PO5511D

Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates for passive and active air sampling applications.

Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.

Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover

  • The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
     
  • Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.

Specifications

Color Ivory, transparent
Content And Storage 2 to 25°C
Format 55mm Contact Plate
Manufacturing Origin Germany
Media Type Tryptone Soya Agar
Product Type Agar Plate
Fill Weight 13.0g ± 0.5g
Packaging Type Triple Wrapped - Irradiated
Quantity 100/Pk.