missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Triple Folding Magnifier, Large, 15X

Catalog No. S01320
Change view
Click to view available options
Magnification:
15x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S01320 15x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Prop 65

Catalog No. S01320 Supplier United Scientific™ Supplier No. MPT038

Prop 65

Sturdy magnifier for use in the classroom.

  • Sturdy plastic frames with 38 mm (1.5 in.) diameter lenses.
  • These versatile magnifiers can magnify objects up to 15x.

Specifications

Material Plastic
Product Type Lens, Magnifying
Size Large
Diameter (Metric) Lens 3.8 cm
Magnification 15x
Diameter (Englis) Lens 1.5 in.