missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Triple Ear Plug Dispenser

Catalog No. 19129482
Change view
Click to view available options
Height (English):
10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
19129482 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19129482 Supplier Brady™ Supplier No. PD244E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acrylic dispensers serve as a visual reminder for employees to wear their ear plugs. Conveniently mount near hazardous work areas to encourage compliance.

  • Each of the three compartments holds up to 50 pair of standard ear plugs
  • Blue acrylic ear plug dispenser tray allows employees three choices for best fit
  • Special bottom tray design prevents overflow and spillage
  • Text legend - WE CARE WHAT YOU WEAR - in white lettering
  • Use on table top or wall mount
  • 10 H x 12.625 W x 6.75 D in.

Specifications

Depth (English) 6.8 in.
Depth (Metric) 17.1 cm
For Use With (Application) Ear Plug Dispenser to be Wall or Surfaced Mounted Near Work Areas Where Hearing Protection is Required.
Height (English) 10 in.
Height (Metric) 25.4 cm
Holds 50 Pairs
Width (Metric) 32 cm
Width (English) 12.6 in.
Product Type Tray/Box Dispenser
Disposable Reusable
Material Acrylic
Mounting Wall Mountable, Free-standing
Show More Show Less