missing translation for 'onlineSavingsMsg'
Learn More

Triphosphopyridine nucleotide, disodium salt, 97+%

CAS: 24292-60-2 | C21H26N7Na2O17P3 | 787.37 g/mol

$880.49 - $880.49

Chemical Identifiers

CAS 24292-60-2
Molecular Formula C21H26N7Na2O17P3
Molecular Weight (g/mol) 787.37
MDL Number MFCD00065390
InChI Key WSDDJLMGYRLUKR-MWLGWILCNA-L
Synonym nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
PubChem CID 131851653
SMILES [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC360565000
SDS View Documents
Thermo Scientific Chemicals
360565000
500 mg Glass bottle
Each for $880.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24292-60-2
787.37
WSDDJLMGYRLUKR-MWLGWILCNA-L
131851653
C21H26N7Na2O17P3
MFCD00065390
nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
[Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

24292-60-2
Authentic
Glass bottle
MFCD00065390
nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
WSDDJLMGYRLUKR-MWLGWILCNA-L
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
131851653
97+%
Triphosphopyridine nucleotide, disodium salt
Yellow
97% min. (enzymatic,on dry subst.)
C21H26N7Na2O17P3
500 mg
(0.05% in water) Clear colorless to light yellow
[Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
787.37
787.4
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 246-129-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only