missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylvinylsilane 93.0+%, TCI America™
$521.72 - $521.72
Chemical Identifiers
| CAS | 18666-68-7 |
|---|---|
| Molecular Formula | C20H18Si |
| Molecular Weight (g/mol) | 286.449 |
| MDL Number | MFCD00051542 |
| InChI Key | OVOIHGSHJGMSMZ-UHFFFAOYSA-N |
| Synonym | triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane |
| PubChem CID | 87747 |
| IUPAC Name | ethenyl(triphenyl)silane |
| SMILES | C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
Chemical Identifiers
| 18666-68-7 | |
| 286.449 | |
| OVOIHGSHJGMSMZ-UHFFFAOYSA-N | |
| 87747 | |
| C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| C20H18Si | |
| MFCD00051542 | |
| triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane | |
| ethenyl(triphenyl)silane |
Specifications
| 18666-68-7 | |
| White-Yellow | |
| C20H18Si | |
| 5 g | |
| OVOIHGSHJGMSMZ-UHFFFAOYSA-N | |
| ethenyl(triphenyl)silane | |
| 87747 | |
| ≥93.0% (GC) | |
| Triphenylvinylsilane |
| 69°C | |
| 190°C | |
| MFCD00051542 | |
| triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane | |
| C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 286.449 | |
| 286.45 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes