missing translation for 'onlineSavingsMsg'
Learn More

Triphenylvinylsilane, 98%

Catalog No. AAA1271306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1271306 5 g
AAA1271314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1271306 Supplier Thermo Scientific Chemicals Supplier No. A1271306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18666-68-7 | C20H18Si | 286.449 g/mol

Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture and heat. Store under inert gas.

Chemical Identifiers

CAS 18666-68-7
Molecular Formula C20H18Si
Molecular Weight (g/mol) 286.449
MDL Number MFCD00051542
InChI Key OVOIHGSHJGMSMZ-UHFFFAOYSA-N
Synonym triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane
PubChem CID 87747
IUPAC Name ethenyl(triphenyl)silane
SMILES C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 68°C to 71°C
Quantity 5 g
Sensitivity Moisture sensitive
Linear Formula (C6H5)3SiCH=CH2
Beilstein 913423
Solubility Information Insoluble in water.
Formula Weight 286.45
Percent Purity 98%
Chemical Name or Material Triphenylvinylsilane
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 242-487-4
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only