missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin fluoride

Catalog No. AA3960003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960003 1 g
AA3960006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3960003 Supplier Thermo Scientific Chemicals Supplier No. 03960003

CAS: 379-52-2 | C18H17FSn | 371.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 379-52-2
Molecular Formula C18H17FSn
Molecular Weight (g/mol) 371.04
MDL Number MFCD00015356
InChI Key SZNZEEYJVCQHCQ-UHFFFAOYSA-N
Synonym triphenyltin fluoride, fentin fluoride, stannane, fluorotriphenyl, biomet 204, tin, fluorotriphenyl, caswell no. 896h, unii-1yp2l952md, fluoro triphenyl stannane, epa pesticide chemical code 083602, triphenylzinnfluorid
PubChem CID 9786
IUPAC Name fluoro(triphenyl)stannane
SMILES F.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >281°C
Quantity 1 g
UN Number UN3146
Formula Weight 368.99
Physical Form Powder
Chemical Name or Material Triphenyltin fluoride
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 206-833-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only