missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin Chloride 95.0+%, TCI America™

Catalog No. t04475g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
T04475G 5 g
1 options

Catalog No. T04475G

Supplier: TCI America T04475G

Only null left

Chemical Identifiers

CAS 639-58-7
Molecular Formula C18H17ClSn
Molecular Weight (g/mol) 387.49
MDL Number MFCD00000519
InChI Key ZDYYCRIXXMWWQC-UHFFFAOYSA-N
Synonym triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
PubChem CID 12540
ChEBI CHEBI:35036
IUPAC Name triphenyltin chloride
SMILES Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
UN Number 3146
Formula Weight 385.48
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenyltin Chloride
EINECSNumber (3)-2598
RTECSNumber WH6860000
TSCA Yes