missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin chloride, 95%

Catalog No. AC317295000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC317295000 500 g Glass bottle
AC317291000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC317295000 Supplier Thermo Scientific Chemicals Supplier No. 317295000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 639-58-7 | C18H17ClSn | 387.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 639-58-7
Molecular Formula C18H17ClSn
Molecular Weight (g/mol) 387.49
MDL Number MFCD00000519
InChI Key ZDYYCRIXXMWWQC-UHFFFAOYSA-N
Synonym triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
PubChem CID 12540
ChEBI CHEBI:35036
IUPAC Name chloro(triphenyl)stannane
SMILES Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 103.0°C to 108.0°C
Color White
Boiling Point 240.0°C (13.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C6H5)3SnCl
Quantity 500 g
Solubility Information Solubility in water: insoluble.
Formula Weight 385.47
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Triphenyltin chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 211-358-4

RUO – Research Use Only