missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin chloride, 95%

Catalog No. AAL1428914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1428914 25 g
AAL1428922 100 g
AAL1428936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1428914 Supplier Thermo Scientific Chemicals Supplier No. L1428914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 639-58-7 | C18H17ClSn | 387.49 g/mol

Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (0.005g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.

Chemical Identifiers

CAS 639-58-7
Molecular Formula C18H17ClSn
Molecular Weight (g/mol) 387.49
MDL Number MFCD00000519
InChI Key ZDYYCRIXXMWWQC-UHFFFAOYSA-N
Synonym triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
PubChem CID 12540
ChEBI CHEBI:35036
IUPAC Name chloro(triphenyl)stannane
SMILES Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 103°C to 106°C
Boiling Point 240°C (13.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Quantity 25 g
UN Number UN3146
Beilstein 524762
Sensitivity Moisture sensitive
Solubility Information Soluble in water (0.005g/L at 20°C).
Formula Weight 385.48
Percent Purity 95%
Chemical Name or Material Triphenyltin chloride
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 211-358-4
RTECSNumber WH6860000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only