missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylsulfonium Bromide 98.0+%, TCI America™
Supplier: TCI America T16091G
Specifications
| Triphenylsulfonium Bromide | |
| 293°C | |
| C18H15BrS | |
| 1 g | |
| C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-] | |
| 343.282 | |
| 343.28 | |
| Crystalline Powder |
| 3353-89-7 | |
| White | |
| MFCD00428762 | |
| VMJFYMAHEGJHFH-UHFFFAOYSA-M | |
| triphenylsulfanium;bromide | |
| 11984010 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3353-89-7 | |
| 343.282 | |
| VMJFYMAHEGJHFH-UHFFFAOYSA-M | |
| triphenylsulfanium;bromide |
| C18H15BrS | |
| MFCD00428762 | |
| 11984010 | |
| C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-] |
Safety and Handling
TSCA : No