missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsulfonium Bromide 98.0+%, TCI America™

Catalog No. T1609100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1609100MG 100 mg
T16091G 1 g
T16095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1609100MG Supplier TCI America Supplier No. T1609100MG

Chemical Identifiers

CAS 3353-89-7
Molecular Formula C18H15BrS
Molecular Weight (g/mol) 343.282
MDL Number MFCD00428762
InChI Key VMJFYMAHEGJHFH-UHFFFAOYSA-M
PubChem CID 11984010
IUPAC Name triphenylsulfanium;bromide
SMILES C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Melting Point 293°C
Color White
Quantity 100 mg
Formula Weight 343.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylsulfonium Bromide