missing translation for 'onlineSavingsMsg'
Learn More

Triphenylsilane 96.0+%, TCI America™

Catalog No. p-7136381
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T06615G 5 g
T066125G 25 g
2 options

Catalog No. T06615G

Supplier: TCI America T06615G

Only null left

Chemical Identifiers

CAS 789-25-3
Molecular Formula C18H15Si
Molecular Weight (g/mol) 259.403
MDL Number MFCD00003003
InChI Key BZLZKLMROPIZSR-UHFFFAOYSA-N
Synonym triphenylsilane, benzene, 1,1',1-silylidynetris, triphenylsilyl, tri phenyl silicon, triphenylsilyl radical, triphenylsilane 5g, 1,1 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-silylidynetrisbenzene; nsc 12565; triphenylhydrosilane
PubChem CID 6327682
IUPAC Name triphenylsilicon
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Boiling Point 184°C
Melting Point 46°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 260.41
Percent Purity ≥96.0% (GC)
Chemical Name or Material Triphenylsilane