missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylmethyl mercaptan, 97%, Thermo Scientific™
$160.91 - $2,481.63
Chemical Identifiers
| CAS | 3695-77-0 |
|---|---|
| Molecular Formula | C19H16S |
| Molecular Weight (g/mol) | 276.39 |
| MDL Number | MFCD00004854 |
| InChI Key | JQZIKLPHXXBMCA-UHFFFAOYSA-N |
| Synonym | triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl |
| PubChem CID | 77281 |
| IUPAC Name | triphenylmethanethiol |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC140410250
|
N/A | 25g | Glass bottle |
Each for $160.91
|
|
||||
|
AC140411000
|
N/A | 100g | Glass bottle |
Each for $521.84
|
|
||||
|
AC140415000
|
N/A | 500g | Glass bottle |
Each for $2,481.63
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C19H16S | |
| MFCD00004854 | |
| triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
| triphenylmethanethiol |
Specifications
| 3695-77-0 | |
| Authentic | |
| C19H16S | |
| MFCD00004854 | |
| triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
| JQZIKLPHXXBMCA-UHFFFAOYSA-N | |
| triphenylmethanethiol | |
| 77281 | |
| 97% |
| 97.5% min. | |
| 96% min. (HPLC) | |
| (C6H5)3CSH | |
| 06, I, 352 | |
| Solubility in water: insoluble. Other solubilities: soluble in toluene | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S | |
| 276.39 | |
| 276.39 | |
| Triphenylmethyl mercaptan |
Safety and Handling
GHS H Statement
Harmful if inhaled.
GHS Signal Word: Warning
EINECSNumber : 223-020-3
RUO – Research Use Only