missing translation for 'onlineSavingsMsg'
Learn More

Triphenylethylene 98.0+%, TCI America™

Catalog No. T183425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T183425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T183425G Supplier TCI America Supplier No. T183425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-72-0
Molecular Formula C20H16
Molecular Weight (g/mol) 256.348
MDL Number MFCD00004765
InChI Key MKYQPGPNVYRMHI-UHFFFAOYSA-N
Synonym triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b
PubChem CID 6025
ChEBI CHEBI:35034
IUPAC Name 1,2-diphenylethenylbenzene
SMILES C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 25 g
Formula Weight 256.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylethylene
RTECSNumber KX4920000
TSCA Yes