missing translation for 'onlineSavingsMsg'
Learn More

Triphenylethylene, 98+%

Catalog No. AAA1324409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324409 10 g
AAA1324418 50 g
AAA1324430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1324409 Supplier Thermo Scientific Chemicals Supplier No. A1324409

CAS: 58-72-0 | C20H16 | 256.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-72-0
Molecular Formula C20H16
Molecular Weight (g/mol) 256.348
MDL Number MFCD00004765
InChI Key MKYQPGPNVYRMHI-UHFFFAOYSA-N
Synonym triphenylethylene, triphenylethene, ethylene, triphenyl, benzilidenediphenylmethane, 1,1,2-triphenylethylene, ethene-1,1,2-triyltribenzene, 1,2-diphenylvinyl benzene, 1,2,2-triphenylethylene, benzene, 1,1',1-1-ethenyl-2-ylidene tris, unii-s4zlz1k74b
PubChem CID 6025
ChEBI CHEBI:35034
IUPAC Name 1,2-diphenylethenylbenzene
SMILES C1=CC=C(C=C1)C=C(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 67°C to 71°C
Boiling Point 219°C to 221°C (14 mmHg)
Quantity 10 g
Beilstein 1867462
Formula Weight 256.35
Percent Purity ≥98%
Chemical Name or Material Triphenylethylene
Hazard Category H302-H373
Precautionary Statement P260-P264b-P270-P301+P312-P314-P330-P501c
EINECSNumber 200-395-1
RTECSNumber KX4920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only