missing translation for 'onlineSavingsMsg'
Learn More

Triphenylchlorosilane 95.0+%, TCI America™

Catalog No. T09395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09395G 5 g
T093925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09395G Supplier TCI America Supplier No. T09395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-86-8
Molecular Formula C18H15ClSi
Molecular Weight (g/mol) 294.85
MDL Number MFCD00000496
InChI Key MNKYQPOFRKPUAE-UHFFFAOYSA-N
Synonym triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl
PubChem CID 6458
IUPAC Name chlorotriphenylsilane
SMILES Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 96°C
Color White
Boiling Point 208°C
Quantity 5 g
UN Number 3261
Formula Weight 294.85
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylchlorosilane
EINECSNumber (3)-2634
RTECSNumber VV2720000
TSCA Yes
Recommended Storage Refrigerator