missing translation for 'onlineSavingsMsg'
Learn More

Triphenylbismuth Carbonate 98.0+%, TCI America™

Catalog No. p-7136982
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18351G 1 g
T18355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18351G Supplier TCI America Supplier No. T18351G

Chemical Identifiers

CAS 47252-14-2
Molecular Formula C19H15BiO3
MDL Number MFCD00209622
Synonym [Carbonato(2-)-O]triphenylbismuth

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 500.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylbismuth Carbonate
TSCA No
Recommended Storage Refrigerator