missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 99+%

Catalog No. p-3758952
Change view
Click to view available options
Quantity:
250 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147672500 250 g Plastic bottle
AC147670025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147672500 Supplier Thermo Scientific Chemicals Supplier No. 147672500
Only null left

CAS: 115-86-6 | C18H15O4P | 326.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Acidity 0.05mg KOH/g max.
Melting Point 47°C to 53°C
Color White
Boiling Point 370°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5O)3P(O)
Quantity 250 g
Beilstein 06, 179
Merck Index 15, 9915
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone
Formula Weight 326.28
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material Triphenyl phosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 204-112-2

RUO – Research Use Only