missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 98%

Catalog No. p-7044765
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0813030 250 g
AAL081300B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0813030 Supplier Thermo Scientific Chemicals Supplier No. L0813030
Only null left

CAS: 115-86-6 | C18H15O4P | 326.29 g/mol

Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point 48°C to 52°C
Boiling Point 247°C to 250°C (11 mmHg)
Flash Point 223°C (433°F)
Odor Odorless
Quantity 250 g
UN Number UN3077
Beilstein 1888236
Merck Index 14,9742
Solubility Information Reacts with water.
Formula Weight 326.29
Percent Purity 98%
Chemical Name or Material Triphenyl phosphate
Show More Show Less
Hazard Category H373
Precautionary Statement P260-P314-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-112-2
RTECSNumber TC8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only