missing translation for 'onlineSavingsMsg'
Learn More

Trioxsalen 98.0+%, TCI America™

Catalog No. p-7137135
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22671G 1 g
T22675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22671G Supplier TCI America Supplier No. T22671G
Only null left

Chemical Identifiers

CAS 3902-71-4
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00005010
InChI Key FMHHVULEAZTJMA-UHFFFAOYSA-N
Synonym trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
PubChem CID 5585
ChEBI CHEBI:28329
IUPAC Name 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
SMILES CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 228.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trioxsalen
RTECSNumber LV1576000
TSCA Yes