missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trioleyl Phosphite (so called) 98.0+%, TCI America™
Supplier: TCI America P0390500ML
Specifications
| Trioleyl Phosphite (so called) | |
| Yellow | |
| MFCD00059452 | |
| Phosphorous Acid Trioleyl Ester | |
| CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC | |
| 833.405 | |
| 833.40 | |
| Liquid |
| 13023-13-7 | |
| C54H105O3P | |
| 500 mL | |
| QULKPMBZSVVAJM-UHFFFAOYSA-N | |
| tris(octadec-9-enyl) phosphite | |
| 54247678 | |
| ≥98.0% (T) |
Chemical Identifiers
| 13023-13-7 | |
| 833.405 | |
| QULKPMBZSVVAJM-UHFFFAOYSA-N | |
| 54247678 | |
| CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC |
| C54H105O3P | |
| MFCD00059452 | |
| Phosphorous Acid Trioleyl Ester | |
| tris(octadec-9-enyl) phosphite |
Safety and Handling
TSCA : No