missing translation for 'onlineSavingsMsg'
Learn More

Trioleyl Phosphite (so called) 98.0+%, TCI America™

Catalog No. P039025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P039025ML 25 mL
P0390500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P039025ML Supplier TCI America Supplier No. P039025ML
Only null left

Chemical Identifiers

CAS 13023-13-7
Molecular Formula C54H105O3P
Molecular Weight (g/mol) 833.405
MDL Number MFCD00059452
InChI Key QULKPMBZSVVAJM-UHFFFAOYSA-N
Synonym Phosphorous Acid Trioleyl Ester
PubChem CID 54247678
IUPAC Name tris(octadec-9-enyl) phosphite
SMILES CCCCCCCCC=CCCCCCCCCOP(OCCCCCCCCC=CCCCCCCCC)OCCCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 833.40
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trioleyl Phosphite (so called)